siRNA Details
| Gene Symbol: 1500004F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI573481.1 | - | 79 | |||
| AK005145.1 | - | 58 | ||||
| AK158492.1 | - | 530 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1500004F05Rik| Gene Symbol: 1500004F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI573481.1 | - | 79 | |||
| AK005145.1 | - | 58 | ||||
| AK158492.1 | - | 530 | ||||