siRNA Details
| Gene Symbol: Tprkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170488.1 | NP_001163959.1 | 6 | 607 | ||
| NM_176842.3 | NP_789812.1 | 5 | 499 | |||
| GenBank mRNA | BC024858.1 | - | 5 | 461 | 5357332 | |
| BC027162.1 | - | 6 | 588 | 4951567 | ||
| BC027413.1 | - | 5 | 486 | 4022441 | ||
| BM935112.1 | - | 6 | 588 | |||
| BM948212.1 | - | 3 | 236 | |||

Tprkb
Genomic Map