siRNA Details
| Gene Symbol: 1810046K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027217.1 | NP_081493.1 | 5 | 814 | ||
| GenBank mRNA | AK007796.1 | - | 5 | 817 | ||
| BC120857.1 | - | 3 | 598 | 40129780 | ||
| BC120859.1 | - | 3 | 598 | 40129782 | ||

1810046K07Rik
Genomic Map