siRNA Details
| Gene Symbol: Krtcap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027221.3 | NP_081497.2 | 5 | 624 | ||
| GenBank mRNA | AK007999.1 | - | 5 | 537 | ||
| BC016516.1 | - | 4 | 734 | 2609442 | ||
| BC027812.1 | - | 5 | 622 | 5251276 | ||
| BC107024.1 | - | 5 | 601 | 40049838 | ||
| BC107025.1 | - | 5 | 601 | 40049842 | ||
| CK129285.1 | - | 5 | 624 | |||

Krtcap3
Genomic Map