siRNA Details
| Gene Symbol: Polr2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027231.1 | NP_081507.1 | 3 | 154 | ||
| GenBank mRNA | AK007913.1 | - | 3 | 169 | ||
| AK165600.1 | - | 3 | 181 | |||
| BC024419.1 | - | 3 | 154 | 4983359 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr2f| Gene Symbol: Polr2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027231.1 | NP_081507.1 | 3 | 154 | ||
| GenBank mRNA | AK007913.1 | - | 3 | 169 | ||
| AK165600.1 | - | 3 | 181 | |||
| BC024419.1 | - | 3 | 154 | 4983359 | ||