siRNA Details
| Gene Symbol: Ttc39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027238.2 | NP_081514.1 | 13 | 1068 | ||
| GenBank mRNA | AK037208.1 | - | 13 | 1069 | ||
| AK049108.1 | - | 13 | 1119 | |||
| AK132655.1 | - | 8 | 519 | |||
| BC151021.1 | - | 13 | 1030 | 9087934 | ||

Ttc39b
Genomic Map