siRNA Details
Gene Symbol: 1810065E05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027239.2 | NP_081515.2 | 4 | 423 | ||
GenBank mRNA | AK007956.1 | - | 4 | 459 | ||
BC026941.1 | - | 4 | 395 | 4024443 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1810065E05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027239.2 | NP_081515.2 | 4 | 423 | ||
GenBank mRNA | AK007956.1 | - | 4 | 459 | ||
BC026941.1 | - | 4 | 395 | 4024443 |