siRNA Details
| Gene Symbol: Thap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145929.1 | NP_001139401.1 | 2 | 277 | ||
| NM_175152.4 | NP_780361.2 | 2 | 277 | |||
| GenBank mRNA | AF357407.1 | - | 1 | 12 | ||
| AK008225.1 | - | 2 | 249 | |||
| AK038184.1 | - | 2 | 251 | |||
| AK050740.1 | - | 2 | 291 | |||
| BC059091.1 | - | 2 | 262 | |||
| BC094395.1 | - | 2 | 258 | 5700290 | ||
| BC115485.1 | - | 2 | 275 | |||
| BC115486.1 | - | 2 | 275 | |||
| BY013572.1 | - | 2 | 277 | |||

Thap3
Genomic Map