siRNA Details
| Gene Symbol: Aunip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081099.1 | NP_001074568.1 | 3 | 651 | ||
| GenBank mRNA | AK011279.1 | - | 3 | 576 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aunip| Gene Symbol: Aunip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081099.1 | NP_001074568.1 | 3 | 651 | ||
| GenBank mRNA | AK011279.1 | - | 3 | 576 | ||