siRNA Details
| Gene Symbol: Cyp2c66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011707.1 | NP_001011707.1 | 8 | 1235 | ||
| GenBank mRNA | AK008512.2 | - | 7 | 1061 | ||
| AY227734.1 | - | 8 | 1235 | |||
| BC147819.1 | - | 8 | 1232 | 9087536 | ||
| BC147824.1 | - | 8 | 1232 | 9087541 | ||

Cyp2c66
Genomic Map