siRNA Details
| Gene Symbol: 2010107G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027251.3 | NP_081527.1 | 3, 4 | 268 | ||
| GenBank mRNA | AK008338.1 | - | 2, 3 | 188 | ||
| AK035688.1 | - | 3, 4 | 269 | |||
| BC024943.1 | - | 3, 4 | 583 | 3989515 | ||

2010107G23Rik
Genomic Map