siRNA Details
| Gene Symbol: FOXO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170931.1 | NP_001164402.1 | 1 | 426 | ||
| NM_005938.3 | NP_005929.2 | 1 | 426 | |||
| GenBank mRNA | AK300142.1 | - | 1 | 372 | ||
| BC106761.2 | - | 1 | 152 | 40025381 | ||
| BP349083.1 | - | 1 | 399 | |||

FOXO4
Genomic Map