siRNA Details
| Gene Symbol: FOXO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170931.1 | NP_001164402.1 | 3 | 1080 | ||
| NM_005938.3 | NP_005929.2 | 2 | 1245 | |||
| XM_011530957.2 | XP_011529259.1 | 2 | 662 | |||
| GenBank mRNA | AF384029.1 | - | 3 | 700 | ||
| AK300142.1 | - | 2 | 1191 | |||
| BC026735.1 | - | 1 | 354 | |||
| BC106761.2 | - | 2 | 971 | 40025381 | ||
| X93996.1 | - | 2 | 1035 | |||

FOXO4
Genomic Map