siRNA Details
| Gene Symbol: Slc25a32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172402.3 | NP_765990.2 | 5 | 827 | ||
| GenBank mRNA | AK031187.1 | - | 5 | 632 | ||
| AK145178.1 | - | 5 | 787 | |||
| AK167501.1 | - | 5 | 637 | |||
| AK170643.1 | - | 5 | 796 | |||
| AK172029.1 | - | 5 | 795 | |||
| AK172597.1 | - | 5 | 740 | |||
| AK187971.1 | - | 4 | 341 | |||
| BC031874.1 | - | 5 | 761 | 5149984 | ||

Slc25a32
Genomic Map