siRNA Details
| Gene Symbol: Zfp715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027264.3 | NP_081540.3 | 5 | 2126 | ||
| GenBank mRNA | BC040201.1 | - | 6 | 2299 | 4951844 | |
| BC040386.1 | - | 4 | 2269 | 3585171 | ||
| BC057313.1 | - | 5 | 2120 | 6827768 | ||

Zfp715
Genomic Map