siRNA Details
| Gene Symbol: Cdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027276.2 | NP_081552.2 | 6 | 569 | ||
| GenBank mRNA | AK147938.1 | - | 6 | 584 | ||
| AK158014.1 | - | 7 | 842 | |||
| AK167965.1 | - | 6 | 584 | |||
| AK168978.1 | - | 6 | 556 | |||
| AK184740.1 | - | 2 | 162 | |||
| AK206352.1 | - | 4 | 272 | |||
| BC026606.1 | - | 6 | 569 | 4976424 | ||

Cdc16
Genomic Map