siRNA Details
| Gene Symbol: Cdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027276.2 | NP_081552.2 | 10 | 996 | ||
| GenBank mRNA | AK147938.1 | - | 10 | 1011 | ||
| AK167965.1 | - | 10 | 1011 | |||
| AK168978.1 | - | 10 | 983 | |||
| AK179999.1 | - | 340 | ||||
| AK189595.1 | - | 5 | 428 | |||
| AK213896.1 | - | 3 | 184 | |||
| BC026606.1 | - | 10 | 996 | 4976424 | ||

Cdc16
Genomic Map