siRNA Details
| Gene Symbol: 1700026L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027283.1 | NP_081559.1 | 464 | |||
| GenBank mRNA | AK006393.1 | - | 4 | 462 | ||
| AK155297.1 | - | 4 | 498 | |||
| AK155408.1 | - | 4 | 465 | |||
| BC055108.1 | - | 464 | 6775232 | |||

1700026L06Rik
Genomic Map