siRNA Details
| Gene Symbol: Znf41-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC096621.1 | - | 2739 | 30464103 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf41-ps| Gene Symbol: Znf41-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC096621.1 | - | 2739 | 30464103 | ||