siRNA Details
| Gene Symbol: Dancr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012278.1 | - | 3 | 476 | ||
| AK045300.1 | - | 1220 | ||||
| AK172090.1 | - | 2 | 979 | |||
| AK172703.1 | - | 1 | 1162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dancr| Gene Symbol: Dancr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012278.1 | - | 3 | 476 | ||
| AK045300.1 | - | 1220 | ||||
| AK172090.1 | - | 2 | 979 | |||
| AK172703.1 | - | 1 | 1162 | |||