siRNA Details
| Gene Symbol: 2610528A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206684.1 | NP_001193613.1 | 3 | 696 | ||
| GenBank mRNA | AK012157.1 | - | 3 | 699 | ||
| BC049685.1 | - | 645 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610528A11Rik| Gene Symbol: 2610528A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206684.1 | NP_001193613.1 | 3 | 696 | ||
| GenBank mRNA | AK012157.1 | - | 3 | 699 | ||
| BC049685.1 | - | 645 | ||||