siRNA Details
| Gene Symbol: Sdr9c7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027301.3 | NP_081577.1 | 3, 4 | 837 | ||
| GenBank mRNA | AK007857.1 | - | 2, 3 | 508 | ||
| AK037184.1 | - | 3, 4 | 855 | |||
| AY044433.1 | - | 3, 4 | 810 | |||
| BC064820.1 | - | 3, 4 | 837 | 30293068 | ||

Sdr9c7
Genomic Map