siRNA Details
| Gene Symbol: Ankrd60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199955.1 | NP_001186884.1 | 1, 2 | 358 | ||
| NM_027303.1 | NP_081579.1 | 1, 2 | 358 | |||
| GenBank mRNA | AK006104.1 | - | 1, 2 | 294 | ||
| AK006546.1 | - | 1, 2 | 300 | |||
| BY706216.1 | - | 1, 2 | 290 | |||

Ankrd60
Genomic Map