siRNA Details
| Gene Symbol: Zdhhc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027306.3 | NP_081582.1 | 1 | 635 | ||
| GenBank mRNA | AK006552.1 | - | 1 | 623 | ||
| AK131706.1 | - | 1 | 609 | |||
| BC049767.1 | - | 1 | 635 | 6774409 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc25| Gene Symbol: Zdhhc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027306.3 | NP_081582.1 | 1 | 635 | ||
| GenBank mRNA | AK006552.1 | - | 1 | 623 | ||
| AK131706.1 | - | 1 | 609 | |||
| BC049767.1 | - | 1 | 635 | 6774409 | ||