siRNA Details
| Gene Symbol: Lysmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027309.2 | NP_081585.2 | 2 | 337 | ||
| GenBank mRNA | BC048545.1 | - | 2 | 326 | 6589154 | |
| DT905233.1 | - | 2 | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lysmd2| Gene Symbol: Lysmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027309.2 | NP_081585.2 | 2 | 337 | ||
| GenBank mRNA | BC048545.1 | - | 2 | 326 | 6589154 | |
| DT905233.1 | - | 2 | 92 | |||