siRNA Details
Gene Symbol: Cysltr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162412.1 | NP_001155884.1 | 5 | 1101 | ||
NM_133720.3 | NP_598481.2 | 6 | 1158 | |||
GenBank mRNA | AF331658.1 | - | 6 | 1117 | ||
AK008997.1 | - | 5 | 1096 | |||
BC132414.1 | - | 2 | 767 | 40130691 | ||
BC132418.1 | - | 2 | 767 | 40130695 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
