siRNA Details
| Gene Symbol: Smc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133786.3 | NP_598547.1 | 11 | 1656 | ||
| GenBank mRNA | AJ534940.2 | - | 10 | 1532 | ||
| AK048650.1 | - | 6 | 869 | |||
| AK088350.1 | - | 11 | 1639 | |||
| AK141703.1 | - | 3 | 294 | |||
| BC002078.1 | - | 6 | 723 | |||
| BC061481.1 | - | 11 | 1631 | 6512832 | ||
| BC062939.1 | - | 11 | 1656 | 6841276 | ||

Smc4
Genomic Map