siRNA Details
| Gene Symbol: Cyp4f16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024442.1 | NP_077762.1 | 2, 3 | 230 | ||
| GenBank mRNA | AF233646.1 | - | 2, 3 | 230 | ||
| AK078557.1 | - | 2, 3 | 291 | |||
| AK146232.1 | - | 2, 3 | 303 | |||
| BC026539.1 | - | 2, 3 | 262 | 4982322 | ||

Cyp4f16
Genomic Map