siRNA Details
| Gene Symbol: Mllt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286158.1 | NP_001273087.1 | 6 | 582 | ||
| NM_027326.3 | NP_081602.3 | 11 | 1981 | |||
| NM_029931.3 | NP_084207.2 | 7 | 654 | |||
| GenBank mRNA | AF333960.1 | - | 10 | 1642 | ||
| AK008707.1 | - | 7 | 616 | |||
| AK019458.1 | - | 6 | 574 | |||
| AK136066.1 | - | 7 | 1215 | |||
| AK147009.1 | - | 7 | 568 | |||
| BC021420.1 | - | 12 | 1820 | 5012671 | ||
| BY713607.1 | - | 6 | 570 | |||
| DV051568.1 | - | 664 | ||||

Mllt3
Genomic Map