siRNA Details
| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081079.1 | NP_001074548.1 | 5, 6 | 665 | ||
| GenBank mRNA | AK008963.1 | - | 2, 3 | 149 | ||
| BC041673.1 | - | 4, 5 | 2047 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogfrl1| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081079.1 | NP_001074548.1 | 5, 6 | 665 | ||
| GenBank mRNA | AK008963.1 | - | 2, 3 | 149 | ||
| BC041673.1 | - | 4, 5 | 2047 | |||