siRNA Details
| Gene Symbol: Cd209g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027343.3 | NP_081619.3 | 7 | 587 | ||
| GenBank mRNA | AK010071.1 | - | 6 | 548 | ||
| AM286437.1 | - | 7 | 590 | |||
| BC118542.1 | - | 1 | 2751 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd209g| Gene Symbol: Cd209g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027343.3 | NP_081619.3 | 7 | 587 | ||
| GenBank mRNA | AK010071.1 | - | 6 | 548 | ||
| AM286437.1 | - | 7 | 590 | |||
| BC118542.1 | - | 1 | 2751 | |||