siRNA Details
| Gene Symbol: Ctsll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027344.3 | NP_081620.2 | 5 | 661 | ||
| GenBank mRNA | AK145382.1 | - | 5 | 665 | ||
| BC147423.1 | - | 4 | 600 | 9056481 | ||
| BC147424.1 | - | 4 | 600 | 9056499 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsll3| Gene Symbol: Ctsll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027344.3 | NP_081620.2 | 5 | 661 | ||
| GenBank mRNA | AK145382.1 | - | 5 | 665 | ||
| BC147423.1 | - | 4 | 600 | 9056481 | ||
| BC147424.1 | - | 4 | 600 | 9056499 | ||