siRNA Details
| Gene Symbol: Ctsll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027344.3 | NP_081620.2 | 6 | 858 | ||
| GenBank mRNA | AK009935.1 | - | 1 | 253 | ||
| AK145382.1 | - | 6 | 862 | |||
| BC147423.1 | - | 5 | 797 | 9056481 | ||
| BC147424.1 | - | 5 | 797 | 9056499 | ||

Ctsll3
Genomic Map