siRNA Details
| Gene Symbol: Med23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166416.1 | NP_001159888.1 | 15 | 1600 | ||
| NM_027347.3 | NP_081623.3 | 14 | 1582 | |||
| GenBank mRNA | AF507918.1 | - | 15 | 1550 | ||
| AK028545.1 | - | 15 | 1613 | |||
| AK042346.1 | - | 15 | 1550 | |||
| AK129312.1 | - | 14 | 1515 | |||
| BC050916.1 | - | 14 | 1521 | 6405418 | ||

Med23
Genomic Map