siRNA Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144801.2 | NP_659050.2 | 5 | 588 | ||
| GenBank mRNA | AK039173.1 | - | 4 | 658 | ||
| AK087636.1 | - | 4 | 793 | |||
| BB666121.1 | - | 5 | 588 | |||
| BC017618.1 | - | 5 | 582 | 4923610 | ||

Tmem143
Genomic Map