siRNA Details
| Gene Symbol: Kif18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197959.2 | NP_932063.2 | 13 | 2354 | ||
| GenBank mRNA | AK013867.1 | - | 2 | 556 | ||
| AK156592.1 | - | 13 | 2324 | |||
| BC023289.1 | - | 7 | 1315 | |||
| BC049272.1 | - | 4 | 888 | |||
| BC057614.1 | - | 13 | 2322 | 6401536 | ||

Kif18b
Genomic Map