siRNA Details
| Gene Symbol: Zfp619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004139.2 | NP_001004139.2 | 4 | 3367 | ||
| GenBank mRNA | BC080693.1 | - | 4 | 3377 | 30432277 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp619| Gene Symbol: Zfp619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004139.2 | NP_001004139.2 | 4 | 3367 | ||
| GenBank mRNA | BC080693.1 | - | 4 | 3377 | 30432277 | |