siRNA Details
| Gene Symbol: Cd2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285905.1 | NP_001272834.1 | 3 | 358 | ||
| NM_001285906.1 | NP_001272835.1 | 3 | 372 | |||
| NM_001285907.1 | NP_001272836.1 | 3 | 500 | |||
| NM_027353.4 | NP_081629.1 | 3 | 352 | |||
| GenBank mRNA | AK010582.1 | - | 3 | 237 | ||
| AK030393.1 | - | 3 | 234 | |||
| AK031694.1 | - | 3 | 266 | |||
| AK034689.1 | - | 3 | 276 | |||
| AK154346.1 | - | 3 | 237 | |||
| AK164102.1 | - | 2 | 216 | |||
| AK167249.1 | - | 3 | 298 | |||
| AK173122.1 | - | 2 | 194 | |||
| AK213302.1 | - | 1 | 63 | |||
| BC006920.1 | - | 3 | 268 | 3601764 | ||
| BC079575.1 | - | 2 | 426 | |||
| BM944739.1 | - | 3 | 282 | |||
 Back To Top
 Back To Top
 Cd2bp2
Cd2bp2 Genomic Map
Genomic Map Back To Top
 Back To Top