siRNA Details
| Gene Symbol: Cd2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285905.1 | NP_001272834.1 | 6 | 1039 | ||
| NM_001285906.1 | NP_001272835.1 | 6 | 1053 | |||
| NM_001285907.1 | NP_001272836.1 | 6 | 1181 | |||
| NM_027353.4 | NP_081629.1 | 6 | 1033 | |||
| GenBank mRNA | AK010582.1 | - | 6 | 918 | ||
| AK030393.1 | - | 6 | 915 | |||
| AK031694.1 | - | 6 | 947 | |||
| AK034689.1 | - | 6 | 957 | |||
| AK154346.1 | - | 6 | 918 | |||
| AK164102.1 | - | 5 | 898 | |||
| AK167249.1 | - | 6 | 979 | |||
| AK173122.1 | - | 5 | 875 | |||
| BC006920.1 | - | 6 | 949 | 3601764 | ||
| BC079575.1 | - | 5 | 1107 | |||

Cd2bp2
Genomic Map