siRNA Details
| Gene Symbol: Rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027355.2 | NP_081631.2 | 1 | 915 | ||
| GenBank mRNA | AK013946.1 | - | 1 | 875 | ||
| AK053769.1 | - | 1 | 918 | |||
| BC046815.1 | - | 1 | 887 | 5702430 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf168| Gene Symbol: Rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027355.2 | NP_081631.2 | 1 | 915 | ||
| GenBank mRNA | AK013946.1 | - | 1 | 875 | ||
| AK053769.1 | - | 1 | 918 | |||
| BC046815.1 | - | 1 | 887 | 5702430 | ||