siRNA Details
| Gene Symbol: Gtf3c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290484.1 | NP_001277413.1 | 2 | 480 | ||
| NM_148928.3 | NP_683730.1 | 2 | 480 | |||
| GenBank mRNA | AK045419.1 | - | 2 | 72 | ||
| AK137359.1 | - | 2 | 469 | |||
| AK147014.1 | - | 2 | 458 | |||
| BC027055.1 | - | 2 | 447 | 5360397 | ||
| BC027247.1 | - | 2 | 390 | 2631223 | ||
| BC039171.1 | - | 2 | 401 | 4500799 | ||
| BQ179286.1 | - | 2 | 480 | |||

Gtf3c5
Genomic Map