siRNA Details
| Gene Symbol: Chp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027363.1 | NP_081639.1 | 4 | 485 | ||
| GenBank mRNA | AK008392.1 | - | 4 | 319 | ||
| BC119202.1 | - | 4 | 485 | 8733951 | ||
| BC119204.1 | - | 4 | 485 | 8733953 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chp2| Gene Symbol: Chp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027363.1 | NP_081639.1 | 4 | 485 | ||
| GenBank mRNA | AK008392.1 | - | 4 | 319 | ||
| BC119202.1 | - | 4 | 485 | 8733951 | ||
| BC119204.1 | - | 4 | 485 | 8733953 | ||