siRNA Details
| Gene Symbol: Mkrn2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101431.1 | NP_001094901.1 | 3, 4 | 517 | ||
| GenBank mRNA | AA833313.1 | - | 3, 4 | 419 | ||
| AK011120.1 | - | 2, 3 | 440 | |||
| AK028760.1 | - | 2, 3 | 390 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mkrn2os| Gene Symbol: Mkrn2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101431.1 | NP_001094901.1 | 3, 4 | 517 | ||
| GenBank mRNA | AA833313.1 | - | 3, 4 | 419 | ||
| AK011120.1 | - | 2, 3 | 440 | |||
| AK028760.1 | - | 2, 3 | 390 | |||