siRNA Details
| Gene Symbol: Mkrn2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101431.1 | NP_001094901.1 | 4 | 617 | ||
| GenBank mRNA | AK011120.1 | - | 3 | 540 | ||
| AK028760.1 | - | 3 | 490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mkrn2os| Gene Symbol: Mkrn2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101431.1 | NP_001094901.1 | 4 | 617 | ||
| GenBank mRNA | AK011120.1 | - | 3 | 540 | ||
| AK028760.1 | - | 3 | 490 | |||