siRNA Details
| Gene Symbol: Afap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027373.2 | NP_081649.1 | 12 | 1722 | ||
| GenBank mRNA | AK129483.1 | - | 12 | 1649 | ||
| AK147706.1 | - | 12 | 1614 | |||
| AK148000.1 | - | 12 | 1577 | |||
| AK161385.1 | - | 12 | 1673 | |||
| BC050814.1 | - | 12 | 1722 | 6773872 | ||

Afap1
Genomic Map