siRNA Details
| Gene Symbol: Hdac8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027382.3 | NP_081658.1 | 5 | 588 | ||
| GenBank mRNA | AK011332.1 | - | 5 | 589 | ||
| AK034511.1 | - | 5 | 554 | |||
| AK041965.1 | - | 5 | 587 | |||
| AK131998.1 | - | 5 | 801 | |||
| BC061257.1 | - | 5 | 560 | 30251620 | ||

Hdac8
Genomic Map