siRNA Details
| Gene Symbol: Ube2cbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027394.2 | NP_081670.1 | 3 | 385 | ||
| GenBank mRNA | AK011446.1 | - | 3 | 402 | ||
| AK049229.1 | - | 3 | 386 | |||
| BC115686.1 | - | 3 | 354 | 40104268 | ||
| BC115687.1 | - | 3 | 353 | 40104269 | ||

Ube2cbp
Genomic Map