siRNA Details
| Gene Symbol: Basp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027395.2 | NP_081671.1 | 2 | 721 | ||
| GenBank mRNA | AK011545.1 | - | 2 | 687 | ||
| AK046868.1 | - | 2 | 696 | |||
| BC063750.1 | - | 2 | 576 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Basp1| Gene Symbol: Basp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027395.2 | NP_081671.1 | 2 | 721 | ||
| GenBank mRNA | AK011545.1 | - | 2 | 687 | ||
| AK046868.1 | - | 2 | 696 | |||
| BC063750.1 | - | 2 | 576 | |||