siRNA Details
| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177608.3 | NP_808276.2 | 18 | 3266 | ||
| GenBank mRNA | AK145135.1 | - | 18 | 3152 | ||
| AK172920.1 | - | 18 | 3133 | |||
| BC141056.1 | - | 18 | 3226 | 9055668 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Secisbp2l| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177608.3 | NP_808276.2 | 18 | 3266 | ||
| GenBank mRNA | AK145135.1 | - | 18 | 3152 | ||
| AK172920.1 | - | 18 | 3133 | |||
| BC141056.1 | - | 18 | 3226 | 9055668 | ||