siRNA Details
| Gene Symbol: Bag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027404.2 | NP_081680.1 | 2 | 1056 | ||
| GenBank mRNA | AK005534.1 | - | 1 | 873 | ||
| AK160246.1 | - | 2 | 1178 | |||
| BC023099.1 | - | 2 | 980 | |||
| BC027827.1 | - | 1 | 92 | |||
| BC037642.1 | - | 2 | 1068 | 5066679 | ||
| BC055762.1 | - | 2 | 976 | |||

Bag5
Genomic Map